Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Cyclopentene-1-acetic acid

Catalog No. C1128525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1128525G 5 g
C11285225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1128525G Supplier Sigma Aldrich Supplier No. C1128525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Cyclopentene-1-acetic acid

Specifications

CAS 13668-61-6
Melting Point 19°C (lit.)
Density 1.047 g/mL (at 25°C (literature))
Boiling Point 93°C to 94°C (2.5 mmHg)
Molecular Formula C7H10O2
Refractive Index n20/D 1.468 (literature)
Linear Formula C5H7CH2CO2H
Quantity 5 g
MDL Number MFCD00001400
Synonym 2-Cyclopenten-1-ylacetic acid
Molecular Weight (g/mol) 126.15
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.