Learn More
Medchemexpress LLC 2-Cyclopentyl-4-(7-((10-(((S)-1-((2S,4R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl) car | 2922675-91-8 | 98.2% | 917.17 | 100 MG
SDP
Catalog No. 5000392687
Encompass_Preferred
CC-3240 (compound 13) is a molecular glue degrader of CaMKK2 based on CC-8977, with an IC50 of 9 nM. It is suitable for laboratory research applications.
  • Molecular glue degrader of CaMKK2
  • Based on CC-8977
  • IC50 of 9 nM
  • For research use only
Catalog No. 50-003-92687 Supplier Medchemexpress LLC Supplier No. HY163064100MG
CC-3240 (compound 13) is a molecular glue degrader of CaMKK2 based on CC-8977, with an IC50 of 9 nM. It is suitable for laboratory research applications.
  • Molecular glue degrader of CaMKK2
  • Based on CC-8977
  • IC50 of 9 nM
  • For research use only