Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-cyclopentylethanamine

Catalog No. CDS008185 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS008185 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS008185 Supplier Sigma Aldrich Supplier No. CDS008185250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,7-octadiyne is a terminal bis-alkyne and participates in the formation of uranium(iv) vinyl complexes
  • It reacts with frustrated lewis pair p(o-tolyl)3/b(c6f5)3 by acetylene c-c coupling to yield the zwitterionic product
  • It undergoes the reductive cyclization during reaction with re2cl4(++-dppm)2 (dppm = ph2pch2pph2) and affords quadruply bonded dirhenium(iii) complex re2cl3(++,++2-c8h7)(++-dppm)2

Specifications

Molecular Formula C7H15N
Linear Formula C7H15N
MDL Number MFCD00058672
Quantity 10 g
Molecular Weight (g/mol) 113.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.