Alfa Aesar™ 2-Cyclopentylethanol, 98+%

Manufacturer: Alfa Aesar™ B2234403

Quantity 1g

 View more versions of this product

Catalog No. AAB2234403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 766-00-7
    Molecular Formula C7H14O
    Formula Weight 114.19
    MDL Number MFCD00019301
    Beilstein 2069860
    Synonym 2-(2-Hydroxyethyl)cyclopentane
    Chemical Name or Material 2-Cyclopentylethanol

    Specifications

    Formula Weight 114.19
    Molecular Formula C7H14O
    MDL Number MFCD00019301
    CAS 766-00-7
    Beilstein 2069860
    Synonym 2-(2-Hydroxyethyl)cyclopentane
    Health Hazard 1 Warning
    Flash Point 70°C (158°F)
    Boiling Point 182°C
    Quantity 1g
    Chemical Name or Material 2-Cyclopentylethanol
    Assay ≥98%

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit