Learn More
Accel Pharmtech Inc 2-CYCLOPROPYL-1H-PYRROLE 5G
SDP
Supplier:  Accel Pharmtech Inc P13305G
Encompass
2-cyclopropyl-1h-pyrrole, cas no.87385-10-2,97% purity
Catalog No. NC1654616
Add to Cart
Request a Quote