Learn More
Medchemexpress LLC 2-Cyclopropylacetic acid | 5239-82-7 | 99.9% | 100.12 | 1 KG
SDP
Catalog No. 5000366401
Encompass_Preferred
2-Cyclopropylacetic acid is a drug intermediate used for the synthesis of various active compounds. This product is for research use only and not intended for patient use.
  • Liquid appearance with a density of 1.172±0.06 g/cm³
  • Colorless to light yellow
  • Ships at room temperature in the continental US; shipping conditions may vary elsewhere
  • Store in pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-66401 Supplier Medchemexpress LLC Supplier No. HYW0106101KG
Add to Cart
Add to Cart
2-Cyclopropylacetic acid is a drug intermediate used for the synthesis of various active compounds. This product is for research use only and not intended for patient use.
  • Liquid appearance with a density of 1.172±0.06 g/cm³
  • Colorless to light yellow
  • Ships at room temperature in the continental US; shipping conditions may vary elsewhere
  • Store in pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month