Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Cyclopropylmethoxyphenylboronic acid pinacol ester

Catalog No. SYX002001G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002001G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002001G Supplier Sigma Aldrich Supplier No. SYX002001G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Electrochemical reduction of 1,5-diiodopentane at carbon electrodes in dmf containing tetramethylammonium perchlorate has been studied by cyclic voltammetry and controlled-potential electrolysis.1,5-diiodopentane has been used:
  • As crosslinking reagent, to investigate the cross-linking layer-by-layer films of bioreducible poly(2-dimethylaminoethyl methacrylate) and DNA
  • In preparation of (iodoalkylaminocarbene)tungsten complexes

Specifications

CAS 1185836-98-9
Molecular Formula C16H23BO3
Linear Formula C16H23BO3
MDL Number MFCD13152006
Quantity 25 g
Molecular Weight (g/mol) 274.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.