Learn More
Medchemexpress LLC HY-114166 10mg Medchemexpress, 2-D08 CAS:144707-18-6 Purity:>98%
SDP
Catalog No. 501969296
Encompass_Preferred
Medchemexpress, HY-114166 10mg 2-D08 CAS:144707-18-6 2-D08 is a cell permeable, mechanistically unique inhibitor of protein SUMOylation. 2-D08 also inhibits Axl with an IC50 of 0.49 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9296 Supplier Medchemexpress LLC Supplier No. HY11416610MG
Medchemexpress, HY-114166 10mg 2-D08 CAS:144707-18-6 2-D08 is a cell permeable, mechanistically unique inhibitor of protein SUMOylation. 2-D08 also inhibits Axl with an IC50 of 0.49 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.