Learn More
Selleck Chemical LLC 2-D08 10mg 144707-18-6 2',3',4'-trihydroxy flavone
SDP
Supplier:  Selleck Chemical LLC S869610MG
Encompass_Preferred
2-D08 (2',3',4'-trihydroxy flavone) is a cell permeable, mechanistically unique inhibitor of protein sumoylation. It is also inhibits Axl, IRAK4, ROS1, MLK4, GSK3?, RET (c-RET), KDR and PI3K? with IC50 values of 0.49, 3.9, 5.3, 9.8, 11, 11, 17 and 35 nM respectively in biochemical assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8125