Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-2-Decanol

Catalog No. 5579861G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-798-61G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-798-61G Supplier Sigma Aldrich Supplier No. 5579861G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-2-Decanol can be used as a chiral inductor in photoelectrocyclization of tropolone methyl ether.

Specifications

CAS 185913-97-7
Molecular Formula C38H30Cl2O2P2
Refractive Index n20/D 1.4340
Linear Formula C38H30Cl2O2P2
MDL Number MFCD03426987
Quantity 250 mg
Synonym (+)-(5,5'-Dichloro-6,6'-dimethoxy-1,1'-biphenyl)-2,2'-diyl-bis(diphenylphosphine); (+)-2,2'-Bis(diphenylphosphino)-5,5'-dichloro-6,6'-dimethoxy-1,1'-biphenyl; (R)-(+)-Cl-MeO-BIPHEP
Molecular Weight (g/mol) 651.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.