Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Decyne 98.0+%, TCI America™
SDP

Catalog No. D11875ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11875ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11875ML Supplier TCI America Supplier No. D11875ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2384-70-5
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00039993
InChI Key RWDDSTHSVISBEA-UHFFFAOYSA-N
PubChem CID 75424
IUPAC Name dec-2-yne
SMILES CCCCCCCC#CC

Specifications

Color Colorless
Boiling Point 84°C
Quantity 5 mL
UN Number 3295
Formula Weight 138.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Decyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.