Learn More
OHAUS 2 Deepwell MicroPlate Foam
Catalog No. 01804340 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-340 Vortex Attachment
1 options
Catalog No. 01-804-340 Supplier OHAUS Supplier No. 83041420
Add to Cart
Add to Cart
Two Extra Deep (1 1/8 or 3 cm) high-density foam inserts to accommodate one standard deepwell microplate.
  • This insert fits into the 6 in. (152 mm) Platform #146-6005-00 attachment for use with the Vortex-Genie 2 mixers and Vortex-Genie Pulse.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Deepwell Microplate Insert for 6-Inch Platform Head