Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Deoxy-2,2-difluoro-D-erythro-pentonic Acid gamma-Lactone 3,5-Dibenzoate 98.0+%, TCI America™
SDP

Catalog No. D42071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42071G 1 g
D42075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42071G Supplier TCI America Supplier No. D42071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122111-01-7
Molecular Formula C19H14F2O6
Molecular Weight (g/mol) 376.312
MDL Number MFCD08458308
InChI Key SHHNEUNVMZNOID-HUUCEWRRSA-N
PubChem CID 9821015
IUPAC Name [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(=O)O2)(F)F)OC(=O)C3=CC=CC=C3

Specifications

Melting Point 123°C
Color White
Quantity 1 g
Formula Weight 376.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Deoxy-2,2-difluoro-D-erythro-pentonic Acid gamma-Lactone 3,5-Dibenzoate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.