Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-deoxy-2,2-difluoro-D-erythro-pentonic acid γ-lactone 3,5-dibenzoate | 122111-01-7 | MFCD08458308 | ≥97.0% | 376.31 | C19H14F2O6 | 5 G
SDP

Catalog No. 5000210575
Encompass_Preferred

2-Deoxy-2,2-difluoro-D-erythro-pentonic acid γ-lactone 3,5-dibenzoate is a chemically synthesized fluorinated carbohydrate derivative used in glycobiology and carbohydrate chemistry research. It is supplied as a white to off-white solid with high reported purity suitable for laboratory applications.

  • Chemical class: Fluorinated carbohydrate γ-lactone.
  • Cas number: 122111-01-7.
  • Chemical formula: C19H14F2O6.
  • Molecular weight: 376.31 g/mol.
  • Purity: ≥97.0%.
  • Appearance: White to off-white solid.
  • Storage: Store at room temperature (solid); in solvent, store at -80°C (2 years) or -20°C (1 year).
  • Available pack sizes: 5 g, 10 g, 25 g, 50 g, 100 g.

Catalog No. 50-002-10575 Supplier Medchemexpress LLC Supplier No. HYW0500495G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Deoxy-2,2-difluoro-D-erythro-pentonic acid γ-lactone 3,5-dibenzoate is a chemically synthesized fluorinated carbohydrate derivative used in glycobiology and carbohydrate chemistry research. It is supplied as a white to off-white solid with high reported purity suitable for laboratory applications.

  • Chemical class: Fluorinated carbohydrate γ-lactone.
  • Cas number: 122111-01-7.
  • Chemical formula: C19H14F2O6.
  • Molecular weight: 376.31 g/mol.
  • Purity: ≥97.0%.
  • Appearance: White to off-white solid.
  • Storage: Store at room temperature (solid); in solvent, store at -80°C (2 years) or -20°C (1 year).
  • Available pack sizes: 5 g, 10 g, 25 g, 50 g, 100 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.