Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxy-2'-fluoro-4-thiouridine | 10212-16-5 | MFCD30570247 | 99.6% | 262.26 g/mol | C9H11FN2O4S | 5 MG
SDP

Catalog No. 5000284094
Encompass_Preferred

2'-Deoxy-2'-fluoro-4-thiouridine is a modified nucleoside for research applications. It is a purine nucleoside analogue reported to inhibit DNA synthesis and induce apoptosis, making it useful in studies of antitumor mechanisms and nucleic acid chemistry. Supplied as a high-purity solid for laboratory use.

  • Purity 99.6%.
  • Molecular weight 262.26 g/mol.
  • Chemical formula C9H11FN2O4S.
  • CAS number 10212-16-5.
  • Typical use research on antitumor mechanisms and nucleic acid studies.
  • Storage store at low temperature, under inert gas when recommended.

Catalog No. 50-002-84094 Supplier Medchemexpress LLC Supplier No. HYW5397545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoro-4-thiouridine is a modified nucleoside for research applications. It is a purine nucleoside analogue reported to inhibit DNA synthesis and induce apoptosis, making it useful in studies of antitumor mechanisms and nucleic acid chemistry. Supplied as a high-purity solid for laboratory use.

  • Purity 99.6%.
  • Molecular weight 262.26 g/mol.
  • Chemical formula C9H11FN2O4S.
  • CAS number 10212-16-5.
  • Typical use research on antitumor mechanisms and nucleic acid studies.
  • Storage store at low temperature, under inert gas when recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.