Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-2'-fluoro-5-iodouridine | 55612-21-0 | 99.5% | 372.09 g/mol | C9H10FIN2O5 | 1 ML
SDP

Catalog No. 5000282546
Encompass_Preferred

2'-Deoxy-2'-fluoro-5-iodouridine (FIRU) is a purine nucleoside analog used in biochemical and cancer research to probe DNA synthesis and nucleoside metabolism. It is supplied in solution and solid forms for laboratory use; certificates of analysis and safety data sheets are available.

  • High purity (99.5%).
  • Molecular weight 372.09 g/mol.
  • Chemical formula C9H10FIN2O5.
  • Available as 10 mM solution in DMSO (1 mL) and solid vials (25-500 mg).
  • Suitable for nucleoside metabolism and antimetabolite studies.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-82546 Supplier Medchemexpress LLC Supplier No. HYW34266410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoro-5-iodouridine (FIRU) is a purine nucleoside analog used in biochemical and cancer research to probe DNA synthesis and nucleoside metabolism. It is supplied in solution and solid forms for laboratory use; certificates of analysis and safety data sheets are available.

  • High purity (99.5%).
  • Molecular weight 372.09 g/mol.
  • Chemical formula C9H10FIN2O5.
  • Available as 10 mM solution in DMSO (1 mL) and solid vials (25-500 mg).
  • Suitable for nucleoside metabolism and antimetabolite studies.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.