Learn More
Medchemexpress LLC 2'-deoxy-2'-fluoro-5-iodouridine | 55612-21-0 | 99.5% | 372.09 g/mol | C9H10FIN2O5 | 25 MG
SDP
Catalog No. 5000282386
Encompass_Preferred
2'-Deoxy-2'-fluoro-5-iodouridine is a halogenated uridine nucleoside analog used as a research reagent in studies of nucleoside metabolism, nucleic acid incorporation, and preclinical antiviral or anticancer research. It is supplied as a solid laboratory reagent for biochemical and cellular experiments.
  • Halogenated uridine analog for nucleoside metabolism and incorporation studies.
  • Reported purity 99.5% by HPLC, suitable for analytical and preparative research.
  • White to off-white powder, supplied in small mass packages for laboratory use.
  • Molecular formula C9H10FIN2O5; molecular weight 372.09 g/mol.
  • Store refrigerated at 2-8 °C to preserve stability.
Catalog No. 50-002-82386 Supplier Medchemexpress LLC Supplier No. HYW34266425MG
2'-Deoxy-2'-fluoro-5-iodouridine is a halogenated uridine nucleoside analog used as a research reagent in studies of nucleoside metabolism, nucleic acid incorporation, and preclinical antiviral or anticancer research. It is supplied as a solid laboratory reagent for biochemical and cellular experiments.
  • Halogenated uridine analog for nucleoside metabolism and incorporation studies.
  • Reported purity 99.5% by HPLC, suitable for analytical and preparative research.
  • White to off-white powder, supplied in small mass packages for laboratory use.
  • Molecular formula C9H10FIN2O5; molecular weight 372.09 g/mol.
  • Store refrigerated at 2-8 °C to preserve stability.