Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxy-2'-fluoro-5-iodouridine | 55612-21-0 | MFCD17214423 | 99.5% | 372.09 g·mol⁻¹ | C9H10FIN2O5 | 100 MG
SDP

Catalog No. 5000282032
Encompass_Preferred

2'-Deoxy-2'-fluoro-5-iodouridine is a fluorinated, iodinated nucleoside analog supplied for research use in studies of nucleoside metabolism and antitumor mechanisms. Supplier information indicates it can inhibit DNA synthesis and induce apoptosis; material is provided as a high-purity solid suitable for biochemical and preclinical investigations.

  • Molecular formula: C9H10FIN2O5.
  • Molecular weight: 372.09 g·mol⁻¹.
  • CAS number: 55612-21-0.
  • High purity suitable for research applications.
  • Appearance: white to off-white solid.
  • Storage: keep sealed, away from moisture and light; refrigerate as recommended.
  • Typical use: biochemical and preclinical antitumor research.

Catalog No. 50-002-82032 Supplier Medchemexpress LLC Supplier No. HYW342664100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoro-5-iodouridine is a fluorinated, iodinated nucleoside analog supplied for research use in studies of nucleoside metabolism and antitumor mechanisms. Supplier information indicates it can inhibit DNA synthesis and induce apoptosis; material is provided as a high-purity solid suitable for biochemical and preclinical investigations.

  • Molecular formula: C9H10FIN2O5.
  • Molecular weight: 372.09 g·mol⁻¹.
  • CAS number: 55612-21-0.
  • High purity suitable for research applications.
  • Appearance: white to off-white solid.
  • Storage: keep sealed, away from moisture and light; refrigerate as recommended.
  • Typical use: biochemical and preclinical antitumor research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.