Learn More
Medchemexpress LLC 2'-deoxy-2'-fluoro-5-iodouridine | 55612-21-0 | MFCD17214423 | 99.5% | 372.09 g/mol | C9H10FIN2O5 | 50 MG
SDP
Catalog No. 5000282387
Encompass_Preferred
2'-Deoxy-2'-fluoro-5-iodouridine is a purine nucleoside analog used in research to study DNA synthesis inhibition and apoptosis in cell and biochemical assays. It is supplied as a solid and as a ready-to-use DMSO solution and is intended for laboratory research use only.
  • High purity (99.46%) suitable for biochemical and cell-based studies.
  • Molecular formula C9H10FIN2O5 and molecular weight 372.09 g/mol.
  • Available as solid powders and 10 mM DMSO solutions for convenient use.
  • Useful as a tool compound in studies of nucleoside metabolism and antitumor activity.
Catalog No. 50-002-82387 Supplier Medchemexpress LLC Supplier No. HYW34266450MG
2'-Deoxy-2'-fluoro-5-iodouridine is a purine nucleoside analog used in research to study DNA synthesis inhibition and apoptosis in cell and biochemical assays. It is supplied as a solid and as a ready-to-use DMSO solution and is intended for laboratory research use only.
  • High purity (99.46%) suitable for biochemical and cell-based studies.
  • Molecular formula C9H10FIN2O5 and molecular weight 372.09 g/mol.
  • Available as solid powders and 10 mM DMSO solutions for convenient use.
  • Useful as a tool compound in studies of nucleoside metabolism and antitumor activity.