Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-2'-fluoro-β-D-arabino-6-azauridine | 908129-24-8 | 96.9% | 247.18 g·mol⁻¹ | C8H10FN3O5 | 10 MG
SDP

Catalog No. 5000286602
Encompass_Preferred

2'-Deoxy-2'-fluoro-β-D-arabino-6-azauridine is a purine nucleoside analogue provided for laboratory research. It is reported to act via inhibition of DNA synthesis and induction of apoptosis and is supplied as a solid suitable for biochemical and cellular studies. Handle according to laboratory safety guidelines and the product safety data sheet.

  • Purity 96.9% suitable for research use.
  • Solid form, off-white to light yellow.
  • Molecular formula C8H10FN3O5.
  • Molecular weight 247.18 g·mol⁻¹.
  • Intended for in vitro research use only; not for human use.

Catalog No. 50-002-86602 Supplier Medchemexpress LLC Supplier No. HY15237810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoro-β-D-arabino-6-azauridine is a purine nucleoside analogue provided for laboratory research. It is reported to act via inhibition of DNA synthesis and induction of apoptosis and is supplied as a solid suitable for biochemical and cellular studies. Handle according to laboratory safety guidelines and the product safety data sheet.

  • Purity 96.9% suitable for research use.
  • Solid form, off-white to light yellow.
  • Molecular formula C8H10FN3O5.
  • Molecular weight 247.18 g·mol⁻¹.
  • Intended for in vitro research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.