Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoro-β-D-arabino-6-azauridine | 908129-24-8 | C8H10FN3O5 | 5 MG
SDP
Catalog No. 5000372599
Encompass_Preferred
2'-Deoxy-2'-fluoro-β-D-arabino-6-azauridine is a purine nucleoside analog that primarily targets DNA synthesis pathways. It exhibits broad antitumor activity and is particularly effective against indolent lymphoid malignancies. Its mechanisms of action include the inhibition of DNA replication and the induction of apoptosis, making it a valuable tool for cancer research and therapeutic development in oncology.
  • Primarily targets DNA synthesis pathways.
  • Exhibits broad antitumor activity.
  • Effective against indolent lymphoid malignancies.
  • Inhibits DNA replication.
  • Induces apoptosis.
  • Valuable tool for cancer research.
  • Useful for therapeutic development in oncology.
Catalog No. 50-003-72599 Supplier Medchemexpress LLC Supplier No. HY1523785MG
2'-Deoxy-2'-fluoro-β-D-arabino-6-azauridine is a purine nucleoside analog that primarily targets DNA synthesis pathways. It exhibits broad antitumor activity and is particularly effective against indolent lymphoid malignancies. Its mechanisms of action include the inhibition of DNA replication and the induction of apoptosis, making it a valuable tool for cancer research and therapeutic development in oncology.
  • Primarily targets DNA synthesis pathways.
  • Exhibits broad antitumor activity.
  • Effective against indolent lymphoid malignancies.
  • Inhibits DNA replication.
  • Induces apoptosis.
  • Valuable tool for cancer research.
  • Useful for therapeutic development in oncology.