Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoro-β-D-arabinocytidine | 56632-83-8 | 99.9% | 245.21 g/mol | C9H12FN3O4 | 1 ML
SDP
Catalog No. 5000282612
Encompass_Preferred
2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated cytidine nucleoside analog provided for biochemical and cell-based research. It is supplied as solid material or as a ready-to-use 10 mM solution in DMSO, and is characterized by a molecular weight of 245.21 g/mol, CAS 56632-83-8, and high reported purity.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Also available as solid in multiple mass sizes for custom preparation.
  • High purity suitable for analytical and biological experiments.
  • Fluorinated nucleoside analog useful for incorporation and enzymatic studies.
  • Documentation provided: datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-82612 Supplier Medchemexpress LLC Supplier No. HYW12878810MM/1ML
2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated cytidine nucleoside analog provided for biochemical and cell-based research. It is supplied as solid material or as a ready-to-use 10 mM solution in DMSO, and is characterized by a molecular weight of 245.21 g/mol, CAS 56632-83-8, and high reported purity.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Also available as solid in multiple mass sizes for custom preparation.
  • High purity suitable for analytical and biological experiments.
  • Fluorinated nucleoside analog useful for incorporation and enzymatic studies.
  • Documentation provided: datasheet, certificate of analysis, and safety data sheet.