Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-2'-fluoro-β-D-arabinocytidine | 56632-83-8 | 99.9% | 245.21 g/mol | C9H12FN3O4 | 100 MG
SDP

Catalog No. 5000282951
Encompass_Preferred

2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated nucleoside analogue used as a research reagent in studies of nucleoside metabolism and antitumor activity. It is provided in high-purity solid and solution formats for laboratory use and is intended for research purposes only.

  • High purity (manufacturer reported 99.92%).
  • CAS registry number 56632-83-8 for unambiguous identification.
  • Molecular formula C9H12FN3O4 and molecular weight 245.21 g/mol.
  • Available in multiple sizes and as DMSO solutions for assay use.
  • Suitable for studies of DNA synthesis inhibition and apoptosis in cancer research.
  • For research use only; not for clinical or human use.

Catalog No. 50-002-82951 Supplier Medchemexpress LLC Supplier No. HYW128788100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated nucleoside analogue used as a research reagent in studies of nucleoside metabolism and antitumor activity. It is provided in high-purity solid and solution formats for laboratory use and is intended for research purposes only.

  • High purity (manufacturer reported 99.92%).
  • CAS registry number 56632-83-8 for unambiguous identification.
  • Molecular formula C9H12FN3O4 and molecular weight 245.21 g/mol.
  • Available in multiple sizes and as DMSO solutions for assay use.
  • Suitable for studies of DNA synthesis inhibition and apoptosis in cancer research.
  • For research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.