Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoro-β-D-arabinocytidine | 56632-83-8 | 99.9% | 245.21 g/mol | C9H12FN3O4 | 250 MG
SDP
Catalog No. 5000282754
Encompass_Preferred
2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated nucleoside analog supplied for laboratory research. It is used in biochemical and medicinal chemistry studies as a modified cytidine analog for nucleoside metabolism, incorporation experiments, and antiviral/anticancer research.
  • Fluorinated deoxy-arabinocytidine nucleoside analog.
  • High purity suitable for research (99.9%).
  • Chemical formula C9H12FN3O4; molecular weight 245.21 g/mol.
  • Available in multiple pack sizes for lab use.
  • Stable when stored at 4°C; protect from light; longer-term storage in solvent at -80°C.
  • Supplied with documentation such as SDS and COA.
Catalog No. 50-002-82754 Supplier Medchemexpress LLC Supplier No. HYW128788250MG
2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated nucleoside analog supplied for laboratory research. It is used in biochemical and medicinal chemistry studies as a modified cytidine analog for nucleoside metabolism, incorporation experiments, and antiviral/anticancer research.
  • Fluorinated deoxy-arabinocytidine nucleoside analog.
  • High purity suitable for research (99.9%).
  • Chemical formula C9H12FN3O4; molecular weight 245.21 g/mol.
  • Available in multiple pack sizes for lab use.
  • Stable when stored at 4°C; protect from light; longer-term storage in solvent at -80°C.
  • Supplied with documentation such as SDS and COA.