Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoro-β-D-arabinocytidine | 56632-83-8 | MFCD07374399 | >97.0% | 245.21 g·mol⁻1 | C9H12FN3O4 | 25 MG
SDP
Catalog No. 5000282755
Encompass_Preferred
2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated nucleoside analogue used in biochemical and preclinical research. It functions as a modified cytidine analogue for studies of DNA synthesis inhibition, nucleic acid chemistry, and investigations of anticancer or antiviral activity. The compound is supplied as a solid for laboratory use and is documented with CAS and molecular formula identifiers.
  • Fluorinated cytidine analogue for studies of DNA synthesis and metabolism.
  • Applicable to cellular and biochemical assays investigating antitumor and antiviral effects.
  • Suitable for milligram-scale laboratory use as a solid reagent.
  • Serves as a building block in oligonucleotide and nucleic acid chemistry research.
  • Documented by CAS and molecular formula for compound traceability.
Catalog No. 50-002-82755 Supplier Medchemexpress LLC Supplier No. HYW12878825MG
2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated nucleoside analogue used in biochemical and preclinical research. It functions as a modified cytidine analogue for studies of DNA synthesis inhibition, nucleic acid chemistry, and investigations of anticancer or antiviral activity. The compound is supplied as a solid for laboratory use and is documented with CAS and molecular formula identifiers.
  • Fluorinated cytidine analogue for studies of DNA synthesis and metabolism.
  • Applicable to cellular and biochemical assays investigating antitumor and antiviral effects.
  • Suitable for milligram-scale laboratory use as a solid reagent.
  • Serves as a building block in oligonucleotide and nucleic acid chemistry research.
  • Documented by CAS and molecular formula for compound traceability.