Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-2'-fluoro-β-D-arabinocytidine | 56632-83-8 | MFCD07374399 | 99.9% | 245.21 g/mol | C9H12FN3O4 | 50 MG
SDP

Catalog No. 5000283437
Encompass_Preferred

2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated deoxycytidine nucleoside analog intended for research into nucleoside analog pharmacology and antitumor mechanisms. It acts by inhibiting DNA synthesis and promoting apoptosis in susceptible cells, and is supplied as a solid for laboratory use with defined storage recommendations.

  • Fluorinated deoxycytidine nucleoside analog.
  • Inhibits DNA synthesis and induces apoptosis.
  • High purity: 99.9%.
  • Molecular formula C9H12FN3O4; molecular weight 245.21 g/mol.
  • Appearance: white to off-white solid.
  • Storage: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple mg sizes, including 50 MG.

Catalog No. 50-002-83437 Supplier Medchemexpress LLC Supplier No. HYW12878850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoro-β-D-arabinocytidine is a fluorinated deoxycytidine nucleoside analog intended for research into nucleoside analog pharmacology and antitumor mechanisms. It acts by inhibiting DNA synthesis and promoting apoptosis in susceptible cells, and is supplied as a solid for laboratory use with defined storage recommendations.

  • Fluorinated deoxycytidine nucleoside analog.
  • Inhibits DNA synthesis and induces apoptosis.
  • High purity: 99.9%.
  • Molecular formula C9H12FN3O4; molecular weight 245.21 g/mol.
  • Appearance: white to off-white solid.
  • Storage: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple mg sizes, including 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.