Learn More
Medchemexpress LLC 2-Deoxy-2-fluoro-L-f 1mg | 70763-62-1 | 166.15 | C6H11FO4 | 1 MG
SDP
Catalog No. 5000308019
Encompass_Preferred
2-Deoxy-2-fluoro-L-fucose is an L-fucose analogue used as a research tool to inhibit cellular fucosylation by interfering with GDP-fucose synthesis; it is commonly employed in studies of glycosylation-dependent processes, including cancer-related pathways.
  • Inhibits fucosylation by interfering with GDP-fucose synthesis.
  • High purity suitable for biochemical and cellular assays.
  • Available in small milligram quantities for research-scale experiments.
  • White to off-white solid, convenient for handling and storage.
  • Supports studies of glycosylation biology and disease-related glycan changes.
Catalog No. 50-003-08019 Supplier Medchemexpress LLC Supplier No. HY1167051MG
2-Deoxy-2-fluoro-L-fucose is an L-fucose analogue used as a research tool to inhibit cellular fucosylation by interfering with GDP-fucose synthesis; it is commonly employed in studies of glycosylation-dependent processes, including cancer-related pathways.
  • Inhibits fucosylation by interfering with GDP-fucose synthesis.
  • High purity suitable for biochemical and cellular assays.
  • Available in small milligram quantities for research-scale experiments.
  • White to off-white solid, convenient for handling and storage.
  • Supports studies of glycosylation biology and disease-related glycan changes.