Learn More
Carbosynth LLC. 2-Deoxy-2-fluoro-L-fucose, 70763-62-1, MFCD08457748, 5 mg
SDP
Supplier:  Carbosynth LLC. MD060895MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Chemical Formula: C6H11FO4, MW: 166.15, Synonyms: 2FF. Can be metabolised inside the cell to a substrate-based inhibitor of fucosyltransferases.
Catalog No. NC1049775