Learn More
Cayman Chemical 2-deoxy-2-fluoro L-Fucose 25mg
SDP
Catalog No. 502966453
Encompass_Preferred
A fluorinated fucose analog that can be metabolized inside the cell to a substrate-based inhibitor of fucosyltransferases
Catalog No. 50-296-6453 Supplier Cayman Chemical Supplier No. 1717125 MG
Add to Cart
Add to Cart
A fluorinated fucose analog that can be metabolized inside the cell to a substrate-based inhibitor of fucosyltransferases