Learn More
Medchemexpress LLC 2′-deoxy-2′-fluoroadenosine | 64183-27-3 | 99.7% | 269.24 g/mol | C10H12FN5O3 | 100 MG
SDP
Catalog No. 5000299813
Encompass_Preferred
2′-Deoxy-2′-fluoroadenosine is a fluorinated deoxyadenosine nucleoside analog used in biochemical research for its antiviral and antitumor activity. It can be incorporated into DNA to disrupt viral or cancer cell replication and is used as a building block for synthesis of 2′-fluoro-modified oligonucleotides. The compound can be enzymatically converted to 2-fluoroadenine, enabling enzyme-directed activation studies.
  • Fluorinated deoxyadenosine nucleoside analog.
  • Interferes with DNA replication in viral and tumor models.
  • Useful for synthesis of 2′-fluoro-modified oligonucleotides.
  • High purity suitable for research applications (≈99.7%).
  • Available in mg-scale pack sizes for laboratory use.
Catalog No. 50-002-99813 Supplier Medchemexpress LLC Supplier No. HYW039442100MG
2′-Deoxy-2′-fluoroadenosine is a fluorinated deoxyadenosine nucleoside analog used in biochemical research for its antiviral and antitumor activity. It can be incorporated into DNA to disrupt viral or cancer cell replication and is used as a building block for synthesis of 2′-fluoro-modified oligonucleotides. The compound can be enzymatically converted to 2-fluoroadenine, enabling enzyme-directed activation studies.
  • Fluorinated deoxyadenosine nucleoside analog.
  • Interferes with DNA replication in viral and tumor models.
  • Useful for synthesis of 2′-fluoro-modified oligonucleotides.
  • High purity suitable for research applications (≈99.7%).
  • Available in mg-scale pack sizes for laboratory use.