Learn More
Medchemexpress LLC 2'-deoxy-2'-fluoroadenosine | 64183-27-3 | MFCD09750859 | 99.7% | 269.24 | C10H12FN5O3 | 1 ML
SDP
Catalog No. 5000299759
Encompass_Preferred
2'-Deoxy-2'-fluoroadenosine is a fluorinated deoxyadenosine nucleoside analog used in biochemical, antiviral, and antitumor research. It is supplied in both solid form and as a 10 mM solution in DMSO to support a range of laboratory applications, and has associated safety and technical documentation available.
Catalog No. 50-002-99759 Supplier Medchemexpress LLC Supplier No. HYW03944210MM/1ML
2'-Deoxy-2'-fluoroadenosine is a fluorinated deoxyadenosine nucleoside analog used in biochemical, antiviral, and antitumor research. It is supplied in both solid form and as a 10 mM solution in DMSO to support a range of laboratory applications, and has associated safety and technical documentation available.