Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-2'-fluoroarabino inosine | 98983-40-5 | 95.0% | 270.22 g·mol⁻¹ | C10H11FN4O4 | 10 MG
SDP

Catalog No. 5000286147
Encompass_Preferred

2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog provided as a white to off-white solid for research use. It is used as a nucleoside antimetabolite in biochemical and medicinal chemistry studies and is supplied with a supplier certificate of analysis.

  • White to off-white solid appearance.
  • Molecular formula C10H11FN4O4.
  • Molecular weight 270.22 g·mol⁻¹.
  • Purity 95.0% (NMR).
  • Storage: 4°C under nitrogen; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Intended for research applications as a nucleoside analog.

Catalog No. 50-002-86147 Supplier Medchemexpress LLC Supplier No. HY15420410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog provided as a white to off-white solid for research use. It is used as a nucleoside antimetabolite in biochemical and medicinal chemistry studies and is supplied with a supplier certificate of analysis.

  • White to off-white solid appearance.
  • Molecular formula C10H11FN4O4.
  • Molecular weight 270.22 g·mol⁻¹.
  • Purity 95.0% (NMR).
  • Storage: 4°C under nitrogen; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Intended for research applications as a nucleoside analog.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.