Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoroarabino inosine | 98983-40-5 | C10H11FN4O4 | 100 MG
SDP
Catalog No. 5000376223
Encompass_Preferred
2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog. These analogs possess broad antitumor activity, specifically targeting indolent lymphoid malignancies. Their anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-76223 Supplier Medchemexpress LLC Supplier No. HY154204100MG
2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog. These analogs possess broad antitumor activity, specifically targeting indolent lymphoid malignancies. Their anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis