Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoroarabino inosine | 98983-40-5 | C10H11FN4O4 | 25 MG
SDP
Catalog No. 5000374978
Encompass_Preferred
2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog with broad antitumor activity targeting indolent lymphoid malignancies. Its anticancer mechanisms involve inhibition of DNA synthesis and induction of apoptosis.
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-74978 Supplier Medchemexpress LLC Supplier No. 25MGHY154204
2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog with broad antitumor activity targeting indolent lymphoid malignancies. Its anticancer mechanisms involve inhibition of DNA synthesis and induction of apoptosis.
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis