Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxy-2'-fluoroarabino inosine | 98983-40-5 | C10H11FN4O4 | 50 MG
SDP

Catalog No. 5000376352
Encompass_Preferred

2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog that exhibits broad antitumor activity. It specifically targets indolent lymphoid malignancies by inhibiting DNA synthesis and inducing apoptosis.

  • Molecular weight: 270.22
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 95.0%
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis

Catalog No. 50-003-76352 Supplier Medchemexpress LLC Supplier No. HY15420450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoroarabino inosine is a purine nucleoside analog that exhibits broad antitumor activity. It specifically targets indolent lymphoid malignancies by inhibiting DNA synthesis and inducing apoptosis.

  • Molecular weight: 270.22
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 95.0%
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.