Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-2'-fluoroarabinoadenosine | 20227-41-2 | MFCD12911815 | >99.0% | 269.23 g/mol | C10H12FN5O3 | 1 ML
SDP

Catalog No. 5000280624
Encompass_Preferred

2'-Deoxy-2'-fluoroarabinoadenosine is a fluorinated nucleoside analogue of deoxyadenosine with reported antitumor and antiviral activity. It is provided for research use as a high-purity biochemical reagent and is commonly supplied as a DMSO solution for biochemical and cell-based assays. The compound is used as a probe in studies of nucleoside metabolism, antiviral screening, and cytotoxicity evaluations.

  • CAS number 20227-41-2.
  • Molecular formula C10H12FN5O3.
  • Molecular weight 269.23 g/mol.
  • Typical purity >99.0% (batch dependent).
  • Supplied as 10 mM solution in DMSO, 1 mL.
  • Form: solid; soluble in DMSO.
  • Applications: antiviral and antitumor research, nucleoside metabolism studies.
  • Storage and handling: follow supplier recommendations and consult the SDS.

Catalog No. 50-002-80624 Supplier Medchemexpress LLC Supplier No. HYW25168710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoroarabinoadenosine is a fluorinated nucleoside analogue of deoxyadenosine with reported antitumor and antiviral activity. It is provided for research use as a high-purity biochemical reagent and is commonly supplied as a DMSO solution for biochemical and cell-based assays. The compound is used as a probe in studies of nucleoside metabolism, antiviral screening, and cytotoxicity evaluations.

  • CAS number 20227-41-2.
  • Molecular formula C10H12FN5O3.
  • Molecular weight 269.23 g/mol.
  • Typical purity >99.0% (batch dependent).
  • Supplied as 10 mM solution in DMSO, 1 mL.
  • Form: solid; soluble in DMSO.
  • Applications: antiviral and antitumor research, nucleoside metabolism studies.
  • Storage and handling: follow supplier recommendations and consult the SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.