Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluorocytidine | 10212-20-1 | 100.0% | 245.21 | 1 G
SDP
Catalog No. 5000452617
Encompass_Preferred
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can synergistically enhance the antiviral effects of T705 against CCHFV replication.
  • Potent inhibitor of CCHFV replication.
  • Acts synergistically with T705 to increase antiviral effects on CCHFV.
  • Exhibits antiviral activities with EC50 of 61 nM against CCHFV.
  • Shows antiviral activities with EC50 of 31 nM against CCHFV/ZsG in Huh7 cells.
  • Has a CC50 of >50.0 μM in Huh7 cells, indicating low cytotoxicity.
Catalog No. 50-004-52617 Supplier Medchemexpress LLC Supplier No. HYW0120091G
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can synergistically enhance the antiviral effects of T705 against CCHFV replication.
  • Potent inhibitor of CCHFV replication.
  • Acts synergistically with T705 to increase antiviral effects on CCHFV.
  • Exhibits antiviral activities with EC50 of 61 nM against CCHFV.
  • Shows antiviral activities with EC50 of 31 nM against CCHFV/ZsG in Huh7 cells.
  • Has a CC50 of >50.0 μM in Huh7 cells, indicating low cytotoxicity.