Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluorocytidine | 10212-20-1 | 99.9% | 245.21 | 100 MG
SDP
Catalog No. 5000452294
Encompass_Preferred
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can work synergistically with T705 to enhance the antiviral effects of both compounds on CCHFV replication.
  • Effectively inhibits Crimean-Congo hemorrhagic fever virus (CCHFV) replication.
  • Can act synergistically with T705 to boost antiviral effects.
  • Intended for research purposes.
  • Available with a purity of 99.98%.
  • Available in solid form and as a 10 mM solution in water.
Catalog No. 50-004-52294 Supplier Medchemexpress LLC Supplier No. HYW012009100MG
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can work synergistically with T705 to enhance the antiviral effects of both compounds on CCHFV replication.
  • Effectively inhibits Crimean-Congo hemorrhagic fever virus (CCHFV) replication.
  • Can act synergistically with T705 to boost antiviral effects.
  • Intended for research purposes.
  • Available with a purity of 99.98%.
  • Available in solid form and as a 10 mM solution in water.