Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluorocytidine | 10212-20-1 | 99.98% | 245.21 | 10 G
SDP
Catalog No. 5000452295
Encompass_Preferred
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can work synergistically with T705 to enhance the antiviral effects of both compounds on CCHFV replication. It has demonstrated antiviral activities with EC50 values of 61 nM and 31 nM against CCHFV and CCHFV/ZsG in Huh7 cells, respectively, and a CC50 of >50.0 μM in Huh7 cells.
  • Potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication.
  • Acts synergistically with T705 to increase antiviral potency.
  • Antiviral activities observed in various cell lines (HEp-2, Huh-7, Vero 76).
  • Available in solid and solution forms.
  • For research use only.
Catalog No. 50-004-52295 Supplier Medchemexpress LLC Supplier No. HYW01200910G
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can work synergistically with T705 to enhance the antiviral effects of both compounds on CCHFV replication. It has demonstrated antiviral activities with EC50 values of 61 nM and 31 nM against CCHFV and CCHFV/ZsG in Huh7 cells, respectively, and a CC50 of >50.0 μM in Huh7 cells.
  • Potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication.
  • Acts synergistically with T705 to increase antiviral potency.
  • Antiviral activities observed in various cell lines (HEp-2, Huh-7, Vero 76).
  • Available in solid and solution forms.
  • For research use only.