Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluorocytidine | 10212-20-1 | 99.98% | 245.21 | 10 MM * 1 ML
SDP
Catalog No. 5000453482
Encompass_Preferred
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It is intended for research use only.
  • Nucleoside analog
  • Potent inhibitor of CCHFV replication
  • Can synergistically increase antiviral potency when combined with another antiviral compound
Catalog No. 50-004-53482 Supplier Medchemexpress LLC Supplier No. HYW01200910MM/1ML
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It is intended for research use only.
  • Nucleoside analog
  • Potent inhibitor of CCHFV replication
  • Can synergistically increase antiviral potency when combined with another antiviral compound