Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluorocytidine | 10212-20-1 | 99.98% | 245.21 | 500 MG
SDP
Catalog No. 5000452785
Encompass_Preferred
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can work synergistically with T705 to enhance the antiviral effects of both compounds on CCHFV replication. It is intended for research use only.
  • Potent inhibitor of CCHFV replication
  • Works synergistically with T705
  • Suitable for research use only
Catalog No. 50-004-52785 Supplier Medchemexpress LLC Supplier No. HYW012009500MG
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can work synergistically with T705 to enhance the antiviral effects of both compounds on CCHFV replication. It is intended for research use only.
  • Potent inhibitor of CCHFV replication
  • Works synergistically with T705
  • Suitable for research use only