Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluorocytidine | 10212-20-1 | MFCD00057445 | 99.98% | 245.21 | 5 G
SDP
Catalog No. 5000452298
Encompass_Preferred
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can also synergistically enhance the antiviral effects of T705 against CCHFV replication.
  • Potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication.
  • Acts synergistically with T705 to increase antiviral effects on CCHFV replication.
  • Available in various quantities and forms: solid, and solutions (e.g., 10 mM * 1 mL in Water).
Catalog No. 50-004-52298 Supplier Medchemexpress LLC Supplier No. HYW0120095G
2'-Deoxy-2'-fluorocytidine is a nucleoside analog that acts as a potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication. It can also synergistically enhance the antiviral effects of T705 against CCHFV replication.
  • Potent inhibitor of Crimean-Congo hemorrhagic fever virus (CCHFV) replication.
  • Acts synergistically with T705 to increase antiviral effects on CCHFV replication.
  • Available in various quantities and forms: solid, and solutions (e.g., 10 mM * 1 mL in Water).