Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoroguanosine | 78842-13-4 | 99.9% | 285.24 | 1 ML
SDP
Catalog No. 5000455028
Encompass_Preferred
2'-Deoxy-2'-fluoroguanosine is a nucleoside analog and a potent inhibitor of the influenza virus, with an EC90 value of <0.35 μM against influenza A and B viruses. It inhibits influenza virus replication in the upper respiratory tract, which helps to alleviate fever and nasal inflammation.
  • Nucleoside analog.
  • Potent influenza virus inhibitor with an EC90 value of <0.35 μM against influenza A and B viruses.
  • Inhibits influenza virus replication in the upper respiratory tract, ameliorating fever and nasal inflammation.
  • For research use only.
Catalog No. 50-004-55028 Supplier Medchemexpress LLC Supplier No. HYW01151810MM/1ML
2'-Deoxy-2'-fluoroguanosine is a nucleoside analog and a potent inhibitor of the influenza virus, with an EC90 value of <0.35 μM against influenza A and B viruses. It inhibits influenza virus replication in the upper respiratory tract, which helps to alleviate fever and nasal inflammation.
  • Nucleoside analog.
  • Potent influenza virus inhibitor with an EC90 value of <0.35 μM against influenza A and B viruses.
  • Inhibits influenza virus replication in the upper respiratory tract, ameliorating fever and nasal inflammation.
  • For research use only.