Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoroinosine | 80049-87-2 | C10H11FN4O4 | 1 G
SDP
Catalog No. 5000358802
Encompass_Preferred
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog. These analogs exhibit broad antitumor activity, particularly against indolent lymphoid malignancies, and their anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Purine nucleoside analog
  • Has broad antitumor activity targeting indolent lymphoid malignancies
  • Anticancer mechanisms include inhibition of DNA synthesis and induction of apoptosis
Catalog No. 50-003-58802 Supplier Medchemexpress LLC Supplier No. HY4829341G
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog. These analogs exhibit broad antitumor activity, particularly against indolent lymphoid malignancies, and their anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Purine nucleoside analog
  • Has broad antitumor activity targeting indolent lymphoid malignancies
  • Anticancer mechanisms include inhibition of DNA synthesis and induction of apoptosis