Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoroinosine | 80049-87-2 | C10H11FN4O4 | 100 MG
SDP
Catalog No. 5000358635
Encompass_Preferred
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog that demonstrates broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms include inhibition of DNA synthesis and induction of apoptosis.
  • Purine nucleoside analog
  • Demonstrates broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Appears as a white to off-white solid
Catalog No. 50-003-58635 Supplier Medchemexpress LLC Supplier No. HY482934100MG
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog that demonstrates broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms include inhibition of DNA synthesis and induction of apoptosis.
  • Purine nucleoside analog
  • Demonstrates broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Appears as a white to off-white solid