Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoroinosine | 80049-87-2 | C10H11FN4O4 | 25 MG
SDP
Catalog No. 5000358934
Encompass_Preferred
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog that exhibits broad antitumor activity against indolent lymphoid malignancies. Its anticancer mechanisms include the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-58934 Supplier Medchemexpress LLC Supplier No. 25MGHY482934
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog that exhibits broad antitumor activity against indolent lymphoid malignancies. Its anticancer mechanisms include the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis