Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluoroinosine | 80049-87-2 | C10H11FN4O4 | 250 MG
SDP
Catalog No. 5000358663
Encompass_Preferred
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Broad antitumor activity targeting indolent lymphoid malignancies.
  • Inhibits DNA synthesis.
  • Induces apoptosis.
Catalog No. 50-003-58663 Supplier Medchemexpress LLC Supplier No. 250MGHY482934
2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Broad antitumor activity targeting indolent lymphoid malignancies.
  • Inhibits DNA synthesis.
  • Induces apoptosis.