Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-2'-fluoroinosine | 80049-87-2 | MFCD09750860 | 270.22 | C10H11FN4O4 | 10 MG
SDP

Catalog No. 5000297870
Encompass_Preferred

2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog used in biomedical research. It exhibits antitumor activity by inhibiting DNA synthesis and inducing apoptosis in preclinical studies. Supplied as a solid powder for laboratory use; consult the manufacturer's data sheet and SDS for handling, purity, and solubility details.

  • Purine nucleoside analog with reported antitumor activity.
  • CAS number 80049-87-2.
  • Molecular formula C10H11FN4O4; molecular weight 270.22.
  • Solid appearance; store powder at -20°C for long-term stability.
  • Supplied in small research pack sizes suitable for in vitro and in vivo studies.

Catalog No. 50-002-97870 Supplier Medchemexpress LLC Supplier No. HY48293410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoroinosine is a purine nucleoside analog used in biomedical research. It exhibits antitumor activity by inhibiting DNA synthesis and inducing apoptosis in preclinical studies. Supplied as a solid powder for laboratory use; consult the manufacturer's data sheet and SDS for handling, purity, and solubility details.

  • Purine nucleoside analog with reported antitumor activity.
  • CAS number 80049-87-2.
  • Molecular formula C10H11FN4O4; molecular weight 270.22.
  • Solid appearance; store powder at -20°C for long-term stability.
  • Supplied in small research pack sizes suitable for in vitro and in vivo studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.