Learn More
Medchemexpress LLC 2'-Deoxy-2'-fluorouridine | 784-71-4 | 99.9% | 246.19 | 1 ML
SDP
Catalog No. 5000454296
Encompass_Preferred
2'-Deoxy-2'-fluorouridine is a derivative of the pyrimidine nucleoside uridine and a nucleoside analog. It inhibits the replication of wild-type viruses by binding to viral RNA. This compound can incorporate into DNA and RNA in rat and woodchuck models upon administration, making it suitable for anti-viral research.
  • Inhibits replication of wild-type viruses.
  • Used in the synthesis of respiratory syncytial virus (RSV) polymerase inhibitors.
  • Suitable for anti-viral research.
Catalog No. 50-004-54296 Supplier Medchemexpress LLC Supplier No. HYW01340310MM/1ML
2'-Deoxy-2'-fluorouridine is a derivative of the pyrimidine nucleoside uridine and a nucleoside analog. It inhibits the replication of wild-type viruses by binding to viral RNA. This compound can incorporate into DNA and RNA in rat and woodchuck models upon administration, making it suitable for anti-viral research.
  • Inhibits replication of wild-type viruses.
  • Used in the synthesis of respiratory syncytial virus (RSV) polymerase inhibitors.
  • Suitable for anti-viral research.