Learn More
Medchemexpress LLC 2′-deoxy-4-thiouridine | 5580-20-1 | 99.8% | 244.27 g·mol⁻¹ | C9H12N2O4S | 10 MG
SDP
Catalog No. 5000280879
Encompass_Preferred
4-Thio-2'-deoxyuridine is a thio-substituted deoxyuridine nucleoside analog used as a probe in nucleic acid chemistry, photochemistry, and cellular studies. The sulfur substitution at C4 gives distinct photophysical and base-pairing properties that facilitate mechanistic, structural, and labeling experiments.
  • High purity (99.8%) suitable for research applications.
  • Available as a solid powder and as a 10 mM solution in DMSO.
  • Offered in multiple package sizes to support small-scale and bulk work.
  • Stable under defined storage conditions for long-term and in-solvent storage.
  • Well characterized with CAS number and molecular formula for unambiguous identification.
Catalog No. 50-002-80879 Supplier Medchemexpress LLC Supplier No. HYW25178110MG
4-Thio-2'-deoxyuridine is a thio-substituted deoxyuridine nucleoside analog used as a probe in nucleic acid chemistry, photochemistry, and cellular studies. The sulfur substitution at C4 gives distinct photophysical and base-pairing properties that facilitate mechanistic, structural, and labeling experiments.
  • High purity (99.8%) suitable for research applications.
  • Available as a solid powder and as a 10 mM solution in DMSO.
  • Offered in multiple package sizes to support small-scale and bulk work.
  • Stable under defined storage conditions for long-term and in-solvent storage.
  • Well characterized with CAS number and molecular formula for unambiguous identification.